Learn More
Grainger G3268 Level Gauge 304 Stainless Steel D 75in
Catalog No. 502766486
Encompass_Preferred
The Krueger 16N553 is a liquid level gauge designed for use with hydraulic reservoirs and gear boxes. It features a polycarbonate lens and aluminum housing for durability. The gauge allows the user to visually check oil level at a glance. It has a maximum pressure rating of 100 PSI and temperature rating of 250°F. The gauge measures 5 inches in diameter and comes with a 1/2 inch NPT bottom mount thread. It can help prevent damage caused by low fluid levels. Overall, the Krueger 16N553 provides a simple, durable liquid level indicator for hydraulic systems.
Catalog No. 50-276-6486 Supplier Grainger Supplier No. 16N553
The Krueger 16N553 is a liquid level gauge designed for use with hydraulic reservoirs and gear boxes. It features a polycarbonate lens and aluminum housing for durability. The gauge allows the user to visually check oil level at a glance. It has a maximum pressure rating of 100 PSI and temperature rating of 250°F. The gauge measures 5 inches in diameter and comes with a 1/2 inch NPT bottom mount thread. It can help prevent damage caused by low fluid levels. Overall, the Krueger 16N553 provides a simple, durable liquid level indicator for hydraulic systems.