Learn More
Grainger G3268 Level Gauge 304 Stainless Steel D 118in
Catalog No. 502766914
Encompass_Preferred
The Krueger 16N593 is a magnetic level gauge that indicates liquid level in tanks and vessels. It has a 316 stainless steel chamber and Teflon float with a magnetic coupling that activates red and green indicator flags outside the chamber as the float rises and falls with the liquid level. This provides a visual display of the tank level through glass windows on the gauge. The unit has flanged connections for installation on tanks up to 15 PSI working pressure from -20 to 250 degrees Fahrenheit. It is designed for use with water, oil and other liquids compatible with stainless steel and Teflon.
Catalog No. 50-276-6914 Supplier Grainger Supplier No. 16N593
The Krueger 16N593 is a magnetic level gauge that indicates liquid level in tanks and vessels. It has a 316 stainless steel chamber and Teflon float with a magnetic coupling that activates red and green indicator flags outside the chamber as the float rises and falls with the liquid level. This provides a visual display of the tank level through glass windows on the gauge. The unit has flanged connections for installation on tanks up to 15 PSI working pressure from -20 to 250 degrees Fahrenheit. It is designed for use with water, oil and other liquids compatible with stainless steel and Teflon.