Learn More
Grainger G3268 Level Gauge 304 Stainless Steel D 49in
Catalog No. 502765725
Encompass_Preferred
The Krueger 16N530 is a liquid level gauge designed for use with hydraulic reservoirs and gear boxes. It features a cast aluminum body with a polycarbonate lens and stainless steel internals. The gauge has a maximum pressure rating of 1000 PSI and temperature rating of 250°F. It uses a bi-color scale to indicate level with green for normal level and red for low level. The gauge has 1/4 NPT bottom mount process connection and is available in 2-1/2", 4" and 5" dial sizes. This rugged and durable gauge allows operators to monitor fluid levels at a glance.
Catalog No. 50-276-5725 Supplier Grainger Supplier No. 16N530
The Krueger 16N530 is a liquid level gauge designed for use with hydraulic reservoirs and gear boxes. It features a cast aluminum body with a polycarbonate lens and stainless steel internals. The gauge has a maximum pressure rating of 1000 PSI and temperature rating of 250°F. It uses a bi-color scale to indicate level with green for normal level and red for low level. The gauge has 1/4 NPT bottom mount process connection and is available in 2-1/2", 4" and 5" dial sizes. This rugged and durable gauge allows operators to monitor fluid levels at a glance.