Learn More
Grainger G3268 Level Gauge 304 Stainless Steel D 85in
Catalog No. 502767107
Encompass_Preferred
The Krueger Level Gauge For 102 in Container is a float-operated mechanical level gauge designed for use in 102 inch high containers. It features a hermetically sealed gauge body and float for use with water-based chemicals. The gauge's scale is calibrated in inches and centimeters for easy fluid level reading. This industrial gauge helps monitor liquid levels in large tanks and vessels. Its stainless steel and polypropylene construction allows compatibility with various corrosive liquids. The Krueger level gauge has a maximum temperature rating of 180°F.
Catalog No. 50-276-7107 Supplier Grainger Supplier No. 16N562
The Krueger Level Gauge For 102 in Container is a float-operated mechanical level gauge designed for use in 102 inch high containers. It features a hermetically sealed gauge body and float for use with water-based chemicals. The gauge's scale is calibrated in inches and centimeters for easy fluid level reading. This industrial gauge helps monitor liquid levels in large tanks and vessels. Its stainless steel and polypropylene construction allows compatibility with various corrosive liquids. The Krueger level gauge has a maximum temperature rating of 180°F.