Learn More
Grainger G3268 Level Gauge 304 Stainless Steel D 119in
Catalog No. 502767433
Encompass_Preferred
The Krueger 16N594 is a liquid level gauge for industrial use, featuring a stainless steel body and float. It indicates liquid level on a 6-inch dial with a 270-degree scale and a maximum pressure rating of 150 PSI. Suitable for various tanks, the oil-filled dial dampens fluctuation. An adjustable pointer kit is included for calibration. The gauge has 1/2 inch NPT bottom and back process connections and requires a minimum liquid specific gravity of 0.5. The Krueger 16N594 offers an analog solution for monitoring liquid levels in industrial settings.
Catalog No. 50-276-7433 Supplier Grainger Supplier No. 16N594
The Krueger 16N594 is a liquid level gauge for industrial use, featuring a stainless steel body and float. It indicates liquid level on a 6-inch dial with a 270-degree scale and a maximum pressure rating of 150 PSI. Suitable for various tanks, the oil-filled dial dampens fluctuation. An adjustable pointer kit is included for calibration. The gauge has 1/2 inch NPT bottom and back process connections and requires a minimum liquid specific gravity of 0.5. The Krueger 16N594 offers an analog solution for monitoring liquid levels in industrial settings.