Learn More
Grainger G3268 Level Gauge 304 Stainless Steel D 29in
Catalog No. 502767595
Encompass_Preferred
The Krueger Level Gauge For 102 in Container is a mechanical level gauge designed for use with 102 inch tall containers. It features a stainless steel body and polypropylene float to provide chemical resistance. The gauge indicates liquid level on a clearly marked scale in 1/2 inch increments. It has 3/4 inch NPT mounting that allows it to be securely installed on the container. This heavy duty mechanical gauge is ideal for use in industrial applications to monitor liquid levels in large tanks and vessels.
Catalog No. 50-276-7595 Supplier Grainger Supplier No. 16N517
The Krueger Level Gauge For 102 in Container is a mechanical level gauge designed for use with 102 inch tall containers. It features a stainless steel body and polypropylene float to provide chemical resistance. The gauge indicates liquid level on a clearly marked scale in 1/2 inch increments. It has 3/4 inch NPT mounting that allows it to be securely installed on the container. This heavy duty mechanical gauge is ideal for use in industrial applications to monitor liquid levels in large tanks and vessels.