Learn More
Grainger G3268 Level Gauge 304 Stainless Steel D 25in
Catalog No. 502767598
Encompass_Preferred
The Krueger 16N513 is a magnetic level gauge designed for use with steam boilers, hot water heating systems, and process tanks to indicate liquid level. This level gauge has a 316 stainless steel chamber with mica viewing windows and an aluminum case. It is made in the USA and is adjustable to fit tank connections from 5-3/4 inches to 7 inches center-to-center. The level gauge comes with Teflon packing nuts and gaskets. It requires no electricity and provides direct reading of liquid level through transparent mica windows. This rugged and reliable magnetic level gauge is ideal for visual monitoring in process applications.
Catalog No. 50-276-7598 Supplier Grainger Supplier No. 16N513
The Krueger 16N513 is a magnetic level gauge designed for use with steam boilers, hot water heating systems, and process tanks to indicate liquid level. This level gauge has a 316 stainless steel chamber with mica viewing windows and an aluminum case. It is made in the USA and is adjustable to fit tank connections from 5-3/4 inches to 7 inches center-to-center. The level gauge comes with Teflon packing nuts and gaskets. It requires no electricity and provides direct reading of liquid level through transparent mica windows. This rugged and reliable magnetic level gauge is ideal for visual monitoring in process applications.