Learn More
Grainger G3268 Level Gauge 304 Stainless Steel D 15in
Catalog No. 502768258
Encompass_Preferred
The Krueger 16N504 is a magnetic level gauge that indicates liquid level in tanks and vessels. It has a 316 stainless steel chamber and Teflon float with a magnetic coupling that activates red and green indicator flags outside the chamber as the float rises and falls with the liquid level. This provides a visual external indication of the liquid level inside the tank. The gauge has flanged ANSI 150 lb connections and is made in the USA. It can withstand temperatures up to 500°F and pressures up to 285 PSI. The gauge comes with a 5 year warranty.
Catalog No. 50-276-8258 Supplier Grainger Supplier No. 16N504
The Krueger 16N504 is a magnetic level gauge that indicates liquid level in tanks and vessels. It has a 316 stainless steel chamber and Teflon float with a magnetic coupling that activates red and green indicator flags outside the chamber as the float rises and falls with the liquid level. This provides a visual external indication of the liquid level inside the tank. The gauge has flanged ANSI 150 lb connections and is made in the USA. It can withstand temperatures up to 500°F and pressures up to 285 PSI. The gauge comes with a 5 year warranty.