Learn More
Grainger G3268 Level Gauge 304 Stainless Steel D 100in
Catalog No. 502768328
Encompass_Preferred
The Krueger 16N576 is a liquid level gauge designed for use in industrial tanks and vessels to visually indicate liquid level. This model features a 10 inch scale length in stainless steel construction with a hermetically sealed glass tube and brass fittings. It has a maximum pressure rating of 30 PSI and temperature rating of 250 degrees Fahrenheit. The gauge provides continuous monitoring of liquid level and can be vertically or horizontally mounted. Overall dimensions are 15 x 2 x 2 inches. This industrial gauge is made in the USA by Krueger Sentry Gauge.
Catalog No. 50-276-8328 Supplier Grainger Supplier No. 16N576
The Krueger 16N576 is a liquid level gauge designed for use in industrial tanks and vessels to visually indicate liquid level. This model features a 10 inch scale length in stainless steel construction with a hermetically sealed glass tube and brass fittings. It has a maximum pressure rating of 30 PSI and temperature rating of 250 degrees Fahrenheit. The gauge provides continuous monitoring of liquid level and can be vertically or horizontally mounted. Overall dimensions are 15 x 2 x 2 inches. This industrial gauge is made in the USA by Krueger Sentry Gauge.