Learn More
Grainger G3268 Level Gauge 304 Stainless Steel D 115in
Catalog No. 502768421
Encompass_Preferred
The Krueger 16N590 is a level gauge designed for use in industrial liquid storage tanks to monitor liquid levels. This model features a 10 inch dial with a 270 degree scale to provide a wide viewing angle. The gauge has a stainless steel stem and brass internals with a maximum pressure rating of 100 PSI. It can accommodate tank depths from 0-36 inches. The gauge comes ready to install with a 1/2 inch NPT lower mount thread connection. This is a mechanical float gauge made in the USA by Krueger Sentry Gauge.
Catalog No. 50-276-8421 Supplier Grainger Supplier No. 16N590
The Krueger 16N590 is a level gauge designed for use in industrial liquid storage tanks to monitor liquid levels. This model features a 10 inch dial with a 270 degree scale to provide a wide viewing angle. The gauge has a stainless steel stem and brass internals with a maximum pressure rating of 100 PSI. It can accommodate tank depths from 0-36 inches. The gauge comes ready to install with a 1/2 inch NPT lower mount thread connection. This is a mechanical float gauge made in the USA by Krueger Sentry Gauge.