Learn More
Grainger G3268 Level Gauge 304 Stainless Steel D 110in
Catalog No. 502768481
Encompass_Preferred
The Krueger 16N585 is a magnetic level gauge for steam boilers, hot water systems, and process tanks, indicating liquid levels visually. It features a 316 stainless steel chamber and a bi-color scale to differentiate water and steam levels. Magnetic couplings allow maintenance without draining. It has a maximum pressure rating of 30 psi at 450°F and a blow-out proof design for safety. With a 10-inch length, it suits residential and commercial boilers. The rugged stainless steel construction ensures durability, making it a high-quality gauge for heating systems and process tanks.
Catalog No. 50-276-8481 Supplier Grainger Supplier No. 16N585
The Krueger 16N585 is a magnetic level gauge for steam boilers, hot water systems, and process tanks, indicating liquid levels visually. It features a 316 stainless steel chamber and a bi-color scale to differentiate water and steam levels. Magnetic couplings allow maintenance without draining. It has a maximum pressure rating of 30 psi at 450°F and a blow-out proof design for safety. With a 10-inch length, it suits residential and commercial boilers. The rugged stainless steel construction ensures durability, making it a high-quality gauge for heating systems and process tanks.