Learn More
Grainger G3269 Level Gauge 304 Stainless Steel D 18in
Catalog No. 502763710
Encompass_Preferred
The Krueger 16N410 is a liquid level gauge designed for use with hydraulic reservoirs and gear boxes. It features a cast aluminum body with a clear polycarbonate sight glass that allows the operator to visually check fluid levels. The gauge has a maximum pressure rating of 1000 PSI and temperature rating of 250°F. It has 1/2" NPT thread connections and comes with a protective rubber boot. The overall dimensions are 2-1/2" diameter x 6-3/4" length. This rugged and durable gauge provides a simple way to monitor fluid levels in industrial equipment.
Catalog No. 50-276-3710 Supplier Grainger Supplier No. 16N410
The Krueger 16N410 is a liquid level gauge designed for use with hydraulic reservoirs and gear boxes. It features a cast aluminum body with a clear polycarbonate sight glass that allows the operator to visually check fluid levels. The gauge has a maximum pressure rating of 1000 PSI and temperature rating of 250°F. It has 1/2" NPT thread connections and comes with a protective rubber boot. The overall dimensions are 2-1/2" diameter x 6-3/4" length. This rugged and durable gauge provides a simple way to monitor fluid levels in industrial equipment.