Learn More
Grainger G3269 Level Gauge 304 Stainless Steel D 57in
Catalog No. 502765139
Encompass_Preferred
The Krueger 16N443 is a liquid level gauge designed for use with hydraulic reservoirs and gear boxes. It features a polycarbonate lens and aluminum housing for durability. The gauge has a dual scale that reads in inches and centimeters with a max level of 8 inches or 20 cm. It uses a bi-color float to indicate level - green for normal level and red when the level drops below 3 inches or 7 cm. The gauge can withstand temperatures from -40 to 257 degrees Fahrenheit. It has a 1/2 inch NPT thread connection and includes a locknut for easy installation on hydraulic reservoirs. The 16N443 provides at-a-glance monitoring of fluid levels.
Catalog No. 50-276-5139 Supplier Grainger Supplier No. 16N443
The Krueger 16N443 is a liquid level gauge designed for use with hydraulic reservoirs and gear boxes. It features a polycarbonate lens and aluminum housing for durability. The gauge has a dual scale that reads in inches and centimeters with a max level of 8 inches or 20 cm. It uses a bi-color float to indicate level - green for normal level and red when the level drops below 3 inches or 7 cm. The gauge can withstand temperatures from -40 to 257 degrees Fahrenheit. It has a 1/2 inch NPT thread connection and includes a locknut for easy installation on hydraulic reservoirs. The 16N443 provides at-a-glance monitoring of fluid levels.