Learn More
Grainger G3269 Level Gauge 304 Stainless Steel D 69in
Catalog No. 502765651
Encompass_Preferred
The Krueger 16N453 is a liquid level gauge designed for use in industrial tanks and vessels to visually indicate liquid level. This model features a 10 inch scale length in aluminum housing with a polycarbonate sight glass. It has brass 1/4 inch NPT process connections and is compatible with water-based liquids. The gauge has a maximum pressure rating of 30 PSI at 150 degrees Fahrenheit. This economical mechanical gauge provides simple, visual monitoring of liquid level.
Catalog No. 50-276-5651 Supplier Grainger Supplier No. 16N453
The Krueger 16N453 is a liquid level gauge designed for use in industrial tanks and vessels to visually indicate liquid level. This model features a 10 inch scale length in aluminum housing with a polycarbonate sight glass. It has brass 1/4 inch NPT process connections and is compatible with water-based liquids. The gauge has a maximum pressure rating of 30 PSI at 150 degrees Fahrenheit. This economical mechanical gauge provides simple, visual monitoring of liquid level.