Learn More
Grainger G3269 Level Gauge 304 Stainless Steel D 89in
Catalog No. 502765728
Encompass_Preferred
The Krueger Level Gauge For 89 in Container is a liquid level gauge designed for 89 inch tall containers. It features a polypropylene float that moves up and down a stainless steel tape to indicate liquid level. The gauge has a maximum reading of 89 inches and minimum of 0 inches, with markings every 1/4 inch. It can handle temperatures up to 180°F. This economical gauge helps monitor liquid levels in large containers.
Catalog No. 50-276-5728 Supplier Grainger Supplier No. 16N471
The Krueger Level Gauge For 89 in Container is a liquid level gauge designed for 89 inch tall containers. It features a polypropylene float that moves up and down a stainless steel tape to indicate liquid level. The gauge has a maximum reading of 89 inches and minimum of 0 inches, with markings every 1/4 inch. It can handle temperatures up to 180°F. This economical gauge helps monitor liquid levels in large containers.