Learn More
Grainger G3269 Level Gauge 304 Stainless Steel D 14in
Catalog No. 502763899
Encompass_Preferred
The Grainger 16N406 is a 16 oz. dead blow plastic hammer with a 2-1/2 inch diameter striking face. It has a hollow head filled with steel and lead shot to prevent rebound and absorb shock. The handle is made of fiberglass reinforced nylon for durability. This hammer allows you to strike delicate surfaces without marring and is well-suited for assembling equipment or driving pins and stakes. It provides the heft of steel with less potential for damage. The hammer weighs 1.1 lbs.
Catalog No. 50-276-3899 Supplier Grainger Supplier No. 16N406
The Grainger 16N406 is a 16 oz. dead blow plastic hammer with a 2-1/2 inch diameter striking face. It has a hollow head filled with steel and lead shot to prevent rebound and absorb shock. The handle is made of fiberglass reinforced nylon for durability. This hammer allows you to strike delicate surfaces without marring and is well-suited for assembling equipment or driving pins and stakes. It provides the heft of steel with less potential for damage. The hammer weighs 1.1 lbs.