Learn More
Grainger G3269 Level Gauge 304 Stainless Steel D 29in
Catalog No. 502763966
Encompass_Preferred
The Krueger 16N420 is a magnetic level gauge that indicates liquid level in tanks and vessels. It has a 316 stainless steel chamber and Teflon float with a magnetic coupling that activates red and green indicator flags outside the chamber as the float rises and falls with the liquid level. This provides a visual display of the liquid level through the glass tube. It has flanged connections for installation on tanks up to 15 PSI working pressure from -20 to 250 degrees Fahrenheit. The gauge has a continuous reading range of 0 to 60 inches.
Catalog No. 50-276-3966 Supplier Grainger Supplier No. 16N420
The Krueger 16N420 is a magnetic level gauge that indicates liquid level in tanks and vessels. It has a 316 stainless steel chamber and Teflon float with a magnetic coupling that activates red and green indicator flags outside the chamber as the float rises and falls with the liquid level. This provides a visual display of the liquid level through the glass tube. It has flanged connections for installation on tanks up to 15 PSI working pressure from -20 to 250 degrees Fahrenheit. The gauge has a continuous reading range of 0 to 60 inches.