Learn More
Grainger G3269 Level Gauge 304 Stainless Steel D 45in
Catalog No. 502767039
Encompass_Preferred
The Grainger 16N432 is a 1/2 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor. It has a NEMA 48Y frame with a 1.15 service factor. The motor is totally enclosed, fan cooled (TEFC) with a cast iron body, aluminum end brackets, and double sealed ball bearings. It can be used for general purpose applications.
Catalog No. 50-276-7039 Supplier Grainger Supplier No. 16N432
The Grainger 16N432 is a 1/2 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor. It has a NEMA 48Y frame with a 1.15 service factor. The motor is totally enclosed, fan cooled (TEFC) with a cast iron body, aluminum end brackets, and double sealed ball bearings. It can be used for general purpose applications.