Learn More
Grainger G3269 Level Gauge 304 Stainless Steel D 32in
Catalog No. 502763711
Encompass_Preferred
The Krueger 16N422 is a liquid level gauge designed for use in industrial tanks and vessels to monitor liquid levels. It features a stainless steel float and stem with a polypropylene indicator. The gauge has a maximum temperature rating of 250°F and a maximum pressure rating of 150 psi. The stem lengths range from 12 inches to 60 inches. This rugged and economical gauge provides continuous monitoring of liquid levels in a variety of applications. With proper installation, it can help prevent tank overfills or running dry. The bright indicator makes reading easy from a distance.
Catalog No. 50-276-3711 Supplier Grainger Supplier No. 16N422
The Krueger 16N422 is a liquid level gauge designed for use in industrial tanks and vessels to monitor liquid levels. It features a stainless steel float and stem with a polypropylene indicator. The gauge has a maximum temperature rating of 250°F and a maximum pressure rating of 150 psi. The stem lengths range from 12 inches to 60 inches. This rugged and economical gauge provides continuous monitoring of liquid levels in a variety of applications. With proper installation, it can help prevent tank overfills or running dry. The bright indicator makes reading easy from a distance.