Learn More
Grainger G3269 Level Gauge 304 Stainless Steel D 87in
Catalog No. 502767249
Encompass_Preferred
The Krueger Level Gauge For 87 in Container is a mechanical level gauge designed for use with 87 inch tall containers. It features a stainless steel body and polypropylene float to provide chemical resistance. The gauge has a 4 inch scale and indicates liquid level from 0 to 87 inches. This mechanical gauge requires no power and is ideal for visual monitoring of liquid levels in stationary tanks and containers up to 87 inches tall. With its durable construction, the Krueger gauge offers a reliable, low-cost solution for monitoring liquid levels.
Catalog No. 50-276-7249 Supplier Grainger Supplier No. 16N469
The Krueger Level Gauge For 87 in Container is a mechanical level gauge designed for use with 87 inch tall containers. It features a stainless steel body and polypropylene float to provide chemical resistance. The gauge has a 4 inch scale and indicates liquid level from 0 to 87 inches. This mechanical gauge requires no power and is ideal for visual monitoring of liquid levels in stationary tanks and containers up to 87 inches tall. With its durable construction, the Krueger gauge offers a reliable, low-cost solution for monitoring liquid levels.