Learn More
Grainger G3269 Level Gauge 304 Stainless Steel D 76in
Catalog No. 502767293
Encompass_Preferred
The Krueger 16N459 is a liquid level gauge designed for industrial applications. It features a stainless steel body and float to withstand harsh environments. The gauge indicates liquid level on a 6 inch dial with a 270 degree scale and has a maximum pressure rating of 150 PSI. This mechanical gauge can be used on a variety of tanks and vessels to monitor liquid levels. The gauge has 1/2 inch NPT thread connections and comes with a mounting bracket for installation. Overall, this is a heavy duty mechanical liquid level gauge suitable for industrial use.
Catalog No. 50-276-7293 Supplier Grainger Supplier No. 16N459
The Krueger 16N459 is a liquid level gauge designed for industrial applications. It features a stainless steel body and float to withstand harsh environments. The gauge indicates liquid level on a 6 inch dial with a 270 degree scale and has a maximum pressure rating of 150 PSI. This mechanical gauge can be used on a variety of tanks and vessels to monitor liquid levels. The gauge has 1/2 inch NPT thread connections and comes with a mounting bracket for installation. Overall, this is a heavy duty mechanical liquid level gauge suitable for industrial use.