Learn More
Grainger G3269 Level Gauge 304 Stainless Steel D 42in
Catalog No. 502767322
Encompass_Preferred
The Krueger 16N430 is a magnetic level gauge designed for use with steam boilers, hot water heating systems, and other pressurized vessels to indicate liquid level. This level gauge has a 316 stainless steel chamber with mica viewing windows and bronze body. It can withstand temperatures up to 750°F and pressures up to 500 psi. The magnetic level indicator provides a direct reading of liquid level through a glass tube against a background of contrasting color. This allows the actual liquid level to be observed instantly at all times. The Krueger 16N430 level gauge offers durability, safety, and easy maintenance.
Catalog No. 50-276-7322 Supplier Grainger Supplier No. 16N430
The Krueger 16N430 is a magnetic level gauge designed for use with steam boilers, hot water heating systems, and other pressurized vessels to indicate liquid level. This level gauge has a 316 stainless steel chamber with mica viewing windows and bronze body. It can withstand temperatures up to 750°F and pressures up to 500 psi. The magnetic level indicator provides a direct reading of liquid level through a glass tube against a background of contrasting color. This allows the actual liquid level to be observed instantly at all times. The Krueger 16N430 level gauge offers durability, safety, and easy maintenance.