Learn More
Grainger G3269 Level Gauge 304 Stainless Steel D 43in
Catalog No. 502767323
Encompass_Preferred
The Krueger Level Gauge For 43 in Container is a mechanical level gauge designed for use with 43 inch diameter containers. It features a stainless steel body and polypropylene float to provide chemical resistance. The gauge indicates liquid level on a clearly marked scale in 1/2 inch increments. It has a maximum reading of 36 inches. This heavy duty gauge is designed for industrial applications. The gauge threads directly into a standard 2 inch opening and is easy to install. It provides accurate monitoring of liquid levels in containers up to 43 inches in diameter.
Catalog No. 50-276-7323 Supplier Grainger Supplier No. 16N431
The Krueger Level Gauge For 43 in Container is a mechanical level gauge designed for use with 43 inch diameter containers. It features a stainless steel body and polypropylene float to provide chemical resistance. The gauge indicates liquid level on a clearly marked scale in 1/2 inch increments. It has a maximum reading of 36 inches. This heavy duty gauge is designed for industrial applications. The gauge threads directly into a standard 2 inch opening and is easy to install. It provides accurate monitoring of liquid levels in containers up to 43 inches in diameter.