Learn More
Grainger G3269 Level Gauge 304 Stainless Steel D 81in
Catalog No. 502767387
Encompass_Preferred
The Krueger 16N464 is a liquid level gauge designed for use in industrial tanks and vessels to visually indicate liquid level. This model features a 10 inch scale length in aluminum housing with a polycarbonate lens. It has a maximum pressure rating of 30 PSI and temperature rating of 250 degrees Fahrenheit. The gauge has 1/4 inch NPT process connections and is compatible with water, oil and other non-corrosive liquids. This is an economical mechanical gauge for basic level indication.
Catalog No. 50-276-7387 Supplier Grainger Supplier No. 16N464
The Krueger 16N464 is a liquid level gauge designed for use in industrial tanks and vessels to visually indicate liquid level. This model features a 10 inch scale length in aluminum housing with a polycarbonate lens. It has a maximum pressure rating of 30 PSI and temperature rating of 250 degrees Fahrenheit. The gauge has 1/4 inch NPT process connections and is compatible with water, oil and other non-corrosive liquids. This is an economical mechanical gauge for basic level indication.