Learn More
Grainger G3269 Level Gauge 304 Stainless Steel D 65in
Catalog No. 502767395
Encompass_Preferred
The Krueger Level Gauge For 65 in Container is a mechanical level gauge designed for use with 65 inch tall containers. It features a stainless steel body with a polypropylene float that moves up and down a center guide rod to indicate liquid level. The gauge has a 6 inch scale and is adjustable to fit containers from 60 to 65 inches tall. It can handle temperatures up to 180°F and pressures up to 100 PSI. This heavy duty mechanical gauge provides accurate level indication without any power required.
Catalog No. 50-276-7395 Supplier Grainger Supplier No. 16N449
The Krueger Level Gauge For 65 in Container is a mechanical level gauge designed for use with 65 inch tall containers. It features a stainless steel body with a polypropylene float that moves up and down a center guide rod to indicate liquid level. The gauge has a 6 inch scale and is adjustable to fit containers from 60 to 65 inches tall. It can handle temperatures up to 180°F and pressures up to 100 PSI. This heavy duty mechanical gauge provides accurate level indication without any power required.