Learn More
Grainger G3269 Level Gauge 304 Stainless Steel D 27in
Catalog No. 502763848
Encompass_Preferred
The Krueger 16N418 is a liquid level gauge designed for use in industrial tanks and vessels to monitor fluid levels. This model features a 10 inch scale length in stainless steel construction with a hermetically sealed magnetic coupling that allows the gauge to be used for a wide range of liquids. The gauge has a dual scale that reads in both inches and centimeters with an accuracy of 1/8 inch. It can withstand operating temperatures from -40 to 250°F and pressures up to 500 PSI. The gauge comes with a 3/4 inch mounting connection. This industrial liquid level gauge provides an accurate visual monitoring solution for fluid levels in processing tanks and vessels.
Catalog No. 50-276-3848 Supplier Grainger Supplier No. 16N418
The Krueger 16N418 is a liquid level gauge designed for use in industrial tanks and vessels to monitor fluid levels. This model features a 10 inch scale length in stainless steel construction with a hermetically sealed magnetic coupling that allows the gauge to be used for a wide range of liquids. The gauge has a dual scale that reads in both inches and centimeters with an accuracy of 1/8 inch. It can withstand operating temperatures from -40 to 250°F and pressures up to 500 PSI. The gauge comes with a 3/4 inch mounting connection. This industrial liquid level gauge provides an accurate visual monitoring solution for fluid levels in processing tanks and vessels.