Learn More
Grainger G3269 Level Gauge 304 Stainless Steel D 51in
Catalog No. 502764916
Encompass_Preferred
The Krueger 16N437 is a liquid level gauge designed for use in industrial tanks and vessels to visually indicate liquid level. This model features a 10 inch clear acrylic sight window protected by a steel frame to allow operators to conveniently monitor liquid level at a glance. The gauge attaches via flange connections and is made of durable steel with a powder coat finish that resists corrosion. With a maximum pressure rating of 15 PSI, this rugged gauge provides reliable visual monitoring for a variety of liquids. The 16N437 is an ideal solution for industrial liquid level indication in processing and storage tanks.
Catalog No. 50-276-4916 Supplier Grainger Supplier No. 16N437
The Krueger 16N437 is a liquid level gauge designed for use in industrial tanks and vessels to visually indicate liquid level. This model features a 10 inch clear acrylic sight window protected by a steel frame to allow operators to conveniently monitor liquid level at a glance. The gauge attaches via flange connections and is made of durable steel with a powder coat finish that resists corrosion. With a maximum pressure rating of 15 PSI, this rugged gauge provides reliable visual monitoring for a variety of liquids. The 16N437 is an ideal solution for industrial liquid level indication in processing and storage tanks.