Learn More
Grainger G3269 Level Gauge 304 Stainless Steel D 37in
Catalog No. 502765002
Encompass_Preferred
The Krueger 16N426 is a magnetic level gauge that indicates liquid level in tanks and vessels. It has a 316 stainless steel chamber and Teflon float with a magnetic coupling that activates red and green indicator flags outside the chamber as the float rises and falls with the liquid level. This provides a visual external indication of the liquid level inside the tank. The gauge has flanged ANSI 150 lb drilling and is suitable for pressures up to 300 PSI at temperatures up to 750°F. It is 30 inches long with a 2 inch diameter. The level gauge offers continuous liquid level monitoring for industrial applications.
Catalog No. 50-276-5002 Supplier Grainger Supplier No. 16N426
The Krueger 16N426 is a magnetic level gauge that indicates liquid level in tanks and vessels. It has a 316 stainless steel chamber and Teflon float with a magnetic coupling that activates red and green indicator flags outside the chamber as the float rises and falls with the liquid level. This provides a visual external indication of the liquid level inside the tank. The gauge has flanged ANSI 150 lb drilling and is suitable for pressures up to 300 PSI at temperatures up to 750°F. It is 30 inches long with a 2 inch diameter. The level gauge offers continuous liquid level monitoring for industrial applications.