Learn More
Grainger G3269 Level Gauge 304 Stainless Steel D 46in
Catalog No. 502765193
Encompass_Preferred
The Krueger 16N433 is a level gauge that provides continuous level indication for liquid level applications. It features a 316 stainless steel float and stem with a hermetically sealed magnetic coupling that actuates the indicator. The gauge has a 6 inch scale and indicates levels from 0 to 12 inches. It can withstand operating temperatures from -40 to 250°F and pressures up to 500 PSI. The gauge has a brass body with a polycarbonate lens and is available with optional materials and configurations for chemical compatibility and hazardous locations. It is designed for use on storage tanks, process vessels, and pipelines in industrial environments.
Catalog No. 50-276-5193 Supplier Grainger Supplier No. 16N433
The Krueger 16N433 is a level gauge that provides continuous level indication for liquid level applications. It features a 316 stainless steel float and stem with a hermetically sealed magnetic coupling that actuates the indicator. The gauge has a 6 inch scale and indicates levels from 0 to 12 inches. It can withstand operating temperatures from -40 to 250°F and pressures up to 500 PSI. The gauge has a brass body with a polycarbonate lens and is available with optional materials and configurations for chemical compatibility and hazardous locations. It is designed for use on storage tanks, process vessels, and pipelines in industrial environments.