Learn More
Grainger G3269 Level Gauge 304 Stainless Steel D 88in
Catalog No. 502765291
Encompass_Preferred
The Krueger Level Gauge For 88 in Container is a mechanical level gauge designed for use with 88 inch tall containers. It features a stainless steel body and polypropylene float to provide chemical resistance. The gauge has a 4 inch dial with gallon graduations and a maximum capacity of 550 gallons. This mechanical gauge allows visual monitoring of liquid level and volume inside the container. Its rugged industrial construction makes it well-suited for use in chemical processing, waste treatment, and other industrial applications requiring sturdy, corrosion-resistant liquid level indication equipment.
Catalog No. 50-276-5291 Supplier Grainger Supplier No. 16N470
The Krueger Level Gauge For 88 in Container is a mechanical level gauge designed for use with 88 inch tall containers. It features a stainless steel body and polypropylene float to provide chemical resistance. The gauge has a 4 inch dial with gallon graduations and a maximum capacity of 550 gallons. This mechanical gauge allows visual monitoring of liquid level and volume inside the container. Its rugged industrial construction makes it well-suited for use in chemical processing, waste treatment, and other industrial applications requiring sturdy, corrosion-resistant liquid level indication equipment.