Learn More
Grainger G3269 Level Gauge 304 Stainless Steel D 68in
Catalog No. 502765292
Encompass_Preferred
The Krueger Level Gauge For 68 in Container is a float-operated mechanical level gauge designed for use with 68 inch tall containers. It features a hermetically sealed reed switch that activates indicator lights to provide a highly visible display of liquid level. This stainless steel gauge has an adjustable specific gravity range from 0.5 to 1.5 and provides continuous level indication in increments of 4 inches. It can handle temperatures up to 250°F and pressures up to 100 PSI. The Krueger level gauge offers durable construction and easy installation.
Catalog No. 50-276-5292 Supplier Grainger Supplier No. 16N452
The Krueger Level Gauge For 68 in Container is a float-operated mechanical level gauge designed for use with 68 inch tall containers. It features a hermetically sealed reed switch that activates indicator lights to provide a highly visible display of liquid level. This stainless steel gauge has an adjustable specific gravity range from 0.5 to 1.5 and provides continuous level indication in increments of 4 inches. It can handle temperatures up to 250°F and pressures up to 100 PSI. The Krueger level gauge offers durable construction and easy installation.