Learn More
Grainger G3269 Level Gauge 304 Stainless Steel D 67in
Catalog No. 502765296
Encompass_Preferred
The Krueger 16N451 is a level gauge that provides continuous level indication for liquid level applications. It features a 316 stainless steel float and stem with a polypropylene indicator. The gauge has a maximum temperature rating of 250°F and a maximum pressure rating of 150 psi. The gauge range is customizable from 6 inches to 60 inches. This rugged and economical gauge is ideal for use with water, diesel fuel, solvents and other liquids in tanks or vessels. With proper installation, it can provide reliable liquid level indication.
Catalog No. 50-276-5296 Supplier Grainger Supplier No. 16N451
The Krueger 16N451 is a level gauge that provides continuous level indication for liquid level applications. It features a 316 stainless steel float and stem with a polypropylene indicator. The gauge has a maximum temperature rating of 250°F and a maximum pressure rating of 150 psi. The gauge range is customizable from 6 inches to 60 inches. This rugged and economical gauge is ideal for use with water, diesel fuel, solvents and other liquids in tanks or vessels. With proper installation, it can provide reliable liquid level indication.