Learn More
Grainger G3269 Level Gauge 304 Stainless Steel D 82in
Catalog No. 502765297
Encompass_Preferred
The Krueger 16N465 is a liquid level gauge designed for industrial applications. It features a stainless steel body and float to withstand harsh environments. The gauge indicates liquid level on a 6 inch dial with a 270 degree scale and has a maximum pressure rating of 150 PSI. This mechanical gauge can be used on a variety of tanks and vessels to monitor liquid levels. The dial is oil-filled to dampen fluctuation. An adjustable pointer kit is included for calibration. The gauge has 1/2 inch NPT process connections and is made in the USA.
Catalog No. 50-276-5297 Supplier Grainger Supplier No. 16N465
The Krueger 16N465 is a liquid level gauge designed for industrial applications. It features a stainless steel body and float to withstand harsh environments. The gauge indicates liquid level on a 6 inch dial with a 270 degree scale and has a maximum pressure rating of 150 PSI. This mechanical gauge can be used on a variety of tanks and vessels to monitor liquid levels. The dial is oil-filled to dampen fluctuation. An adjustable pointer kit is included for calibration. The gauge has 1/2 inch NPT process connections and is made in the USA.