Learn More
Grainger G3269 Level Gauge 304 Stainless Steel D 92in
Catalog No. 502765535
Encompass_Preferred
The Krueger 92-inch Level Gauge is a liquid level indicator designed for 92-inch containers. It features a polypropylene float that moves up and down a polycarbonate indicator tube as liquid level changes. The tube is marked with gallon increments from 0 to 550 gallons. This gauge can withstand temperatures up to 150°F and pressures up to 100 PSI. It has 3/4 inch NPT thread connections and is made in the USA. The overall length is 96 inches. This gauge allows visual monitoring of liquid level in large containers.
Catalog No. 50-276-5535 Supplier Grainger Supplier No. 16N474
The Krueger 92-inch Level Gauge is a liquid level indicator designed for 92-inch containers. It features a polypropylene float that moves up and down a polycarbonate indicator tube as liquid level changes. The tube is marked with gallon increments from 0 to 550 gallons. This gauge can withstand temperatures up to 150°F and pressures up to 100 PSI. It has 3/4 inch NPT thread connections and is made in the USA. The overall length is 96 inches. This gauge allows visual monitoring of liquid level in large containers.