Learn More
Grainger G3269 Level Gauge 304 Stainless Steel D 28in
Catalog No. 502763849
Encompass_Preferred
The Krueger 16N419 is a liquid level gauge designed for use with hydraulic reservoirs and gear boxes. It features a clear acrylic window with green and red zones for quick visual monitoring of oil level. The gauge has a 1/2 inch NPT thread and is made from durable brass construction. It has a maximum pressure rating of 1000 PSI and temperature rating of 250 degrees Fahrenheit. The Krueger 16N419 allows for convenient monitoring of oil level without having to open the reservoir.
Catalog No. 50-276-3849 Supplier Grainger Supplier No. 16N419
The Krueger 16N419 is a liquid level gauge designed for use with hydraulic reservoirs and gear boxes. It features a clear acrylic window with green and red zones for quick visual monitoring of oil level. The gauge has a 1/2 inch NPT thread and is made from durable brass construction. It has a maximum pressure rating of 1000 PSI and temperature rating of 250 degrees Fahrenheit. The Krueger 16N419 allows for convenient monitoring of oil level without having to open the reservoir.