Learn More
Grainger G3269 Level Gauge 304 Stainless Steel D 106in
Catalog No. 502765730
Encompass_Preferred
The Krueger 16N487 is a liquid level gauge designed for use with hydraulic reservoirs and gear boxes. It features a polycarbonate lens and aluminum housing for durability. The gauge allows the user to visually check oil level at a glance. It has a maximum pressure rating of 100 PSI and temperature rating of 250°F. The gauge measures 5 inches in diameter and comes with a 1/2 inch NPT bottom mount thread. This heavy duty gauge is made in the USA by Krueger Sentry Gauge.
Catalog No. 50-276-5730 Supplier Grainger Supplier No. 16N487
The Krueger 16N487 is a liquid level gauge designed for use with hydraulic reservoirs and gear boxes. It features a polycarbonate lens and aluminum housing for durability. The gauge allows the user to visually check oil level at a glance. It has a maximum pressure rating of 100 PSI and temperature rating of 250°F. The gauge measures 5 inches in diameter and comes with a 1/2 inch NPT bottom mount thread. This heavy duty gauge is made in the USA by Krueger Sentry Gauge.