Learn More
Grainger G3269 Level Gauge 304 Stainless Steel D 110in
Catalog No. 502765734
Encompass_Preferred
The Krueger 16N490 is a liquid level gauge designed for use with hydraulic reservoirs and gear boxes. It features a polycarbonate lens for impact and chemical resistance. The gauge has a dual scale that measures in inches and centimeters with a 6 inch range. It has a 1/2 inch NPT bottom connection and includes a locknut for easy installation. The gauge body is made of steel with a baked powder coat finish for durability. This economical gauge allows the user to monitor fluid levels at a glance.
Catalog No. 50-276-5734 Supplier Grainger Supplier No. 16N490
The Krueger 16N490 is a liquid level gauge designed for use with hydraulic reservoirs and gear boxes. It features a polycarbonate lens for impact and chemical resistance. The gauge has a dual scale that measures in inches and centimeters with a 6 inch range. It has a 1/2 inch NPT bottom connection and includes a locknut for easy installation. The gauge body is made of steel with a baked powder coat finish for durability. This economical gauge allows the user to monitor fluid levels at a glance.