Learn More
Grainger G3269 Level Gauge 304 Stainless Steel D 112in
Catalog No. 502768287
Encompass_Preferred
The Krueger 16N492 is a liquid level gauge designed for use in industrial tanks and vessels to visually indicate liquid level. This model features a 10 inch scale length in aluminum housing with a polycarbonate lens. It has a maximum pressure rating of 30 PSI and temperature rating of 250°F. The gauge has 1/4 inch NPT process connections and is compatible with water-based liquids. This economical mechanical gauge provides simple, visual monitoring of liquid level in a range of industrial applications.
Catalog No. 50-276-8287 Supplier Grainger Supplier No. 16N492
The Krueger 16N492 is a liquid level gauge designed for use in industrial tanks and vessels to visually indicate liquid level. This model features a 10 inch scale length in aluminum housing with a polycarbonate lens. It has a maximum pressure rating of 30 PSI and temperature rating of 250°F. The gauge has 1/4 inch NPT process connections and is compatible with water-based liquids. This economical mechanical gauge provides simple, visual monitoring of liquid level in a range of industrial applications.