Learn More
Grainger G3269 Level Gauge 304 Stainless Steel D 53in
Catalog No. 502768448
Encompass_Preferred
The Krueger 16N439 is a level gauge that provides continuous level indication for liquid level applications. It features a 316 stainless steel float and stem with a hermetically sealed magnetic coupling that activates the indicator. The gauge has a 6 inch scale length and indicates levels from 0-12 inches. It has a maximum temperature rating of 250°F and a maximum pressure rating of 150 PSI. The gauge can be mounted to a tank using a 3/4 inch NPT connection. It is made in the USA by Krueger Sentry Gauge.
Catalog No. 50-276-8448 Supplier Grainger Supplier No. 16N439
The Krueger 16N439 is a level gauge that provides continuous level indication for liquid level applications. It features a 316 stainless steel float and stem with a hermetically sealed magnetic coupling that activates the indicator. The gauge has a 6 inch scale length and indicates levels from 0-12 inches. It has a maximum temperature rating of 250°F and a maximum pressure rating of 150 PSI. The gauge can be mounted to a tank using a 3/4 inch NPT connection. It is made in the USA by Krueger Sentry Gauge.