Learn More
Grainger G3269 Level Gauge 304 Stainless Steel D 56in
Catalog No. 502768518
Encompass_Preferred
The Krueger 16N442 is a liquid level gauge designed for use in industrial tanks and vessels to monitor liquid levels. This model features a 10 inch scale length in stainless steel construction with a hermetically sealed gauge glass for viewing fluid levels. It has 150 PSI maximum pressure at 100 degrees F temperature rating. The connections are 1/2 inch NPT thread. This industrial liquid level gauge provides an accurate visual monitoring of tank levels for various liquids. With proper installation, it allows operators to easily check fill levels and prevent overfills.
Catalog No. 50-276-8518 Supplier Grainger Supplier No. 16N442
The Krueger 16N442 is a liquid level gauge designed for use in industrial tanks and vessels to monitor liquid levels. This model features a 10 inch scale length in stainless steel construction with a hermetically sealed gauge glass for viewing fluid levels. It has 150 PSI maximum pressure at 100 degrees F temperature rating. The connections are 1/2 inch NPT thread. This industrial liquid level gauge provides an accurate visual monitoring of tank levels for various liquids. With proper installation, it allows operators to easily check fill levels and prevent overfills.