Learn More
Grainger G3269 Level Gauge 304 Stainless Steel D 19in
Catalog No. 502763697
Encompass_Preferred
The Krueger 16N411 is a liquid level gauge designed for use with hydraulic reservoirs and gear boxes. It features a polycarbonate lens and aluminum housing for durability. The gauge has a dual scale that reads in inches and centimeters with a max level of 8 inches or 20 cm. It uses a bi-color float to indicate level - green for normal level and red when the level drops below minimum. The gauge can withstand temperatures from -40 to 257 degrees Fahrenheit. It has a 1/2 inch NPT thread connection and comes with a locknut for easy install. The Krueger 16N411 provides an easy visual identification of fluid level status.
Catalog No. 50-276-3697 Supplier Grainger Supplier No. 16N411
The Krueger 16N411 is a liquid level gauge designed for use with hydraulic reservoirs and gear boxes. It features a polycarbonate lens and aluminum housing for durability. The gauge has a dual scale that reads in inches and centimeters with a max level of 8 inches or 20 cm. It uses a bi-color float to indicate level - green for normal level and red when the level drops below minimum. The gauge can withstand temperatures from -40 to 257 degrees Fahrenheit. It has a 1/2 inch NPT thread connection and comes with a locknut for easy install. The Krueger 16N411 provides an easy visual identification of fluid level status.