Learn More
Grainger G3269 Level Gauge 304 Stainless Steel D 77in
Catalog No. 502767364
Encompass_Preferred
The Krueger 16N460 is a liquid level gauge designed for use in industrial tanks and vessels to visually indicate liquid level. This model features a 10 inch scale length in aluminum housing with a polycarbonate lens. It has a maximum pressure rating of 30 PSI and temperature rating of 250°F. The gauge has 1/4 inch NPT process connections and is compatible with water, oil and mild chemicals. This direct reading gauge allows operators to conveniently monitor liquid level at a glance.
Catalog No. 50-276-7364 Supplier Grainger Supplier No. 16N460
The Krueger 16N460 is a liquid level gauge designed for use in industrial tanks and vessels to visually indicate liquid level. This model features a 10 inch scale length in aluminum housing with a polycarbonate lens. It has a maximum pressure rating of 30 PSI and temperature rating of 250°F. The gauge has 1/4 inch NPT process connections and is compatible with water, oil and mild chemicals. This direct reading gauge allows operators to conveniently monitor liquid level at a glance.