Learn More
Grainger G3269 Level Gauge 304 Stainless Steel D 12in
Catalog No. 502763979
Encompass_Preferred
The Krueger 16N404 is a liquid level gauge designed for industrial applications. It features a stainless steel float and stem with a polypropylene indicator. The gauge has a maximum temperature rating of 250°F and a maximum pressure rating of 150 psi. It has a 4 inch dial with a dual scale calibrated in inches and centimeters. The gauge provides continuous monitoring of liquid level in tanks or vessels. It can be mounted either vertically or horizontally. This industrial liquid level gauge is made in the USA by Krueger Sentry Gauge.
Catalog No. 50-276-3979 Supplier Grainger Supplier No. 16N404
The Krueger 16N404 is a liquid level gauge designed for industrial applications. It features a stainless steel float and stem with a polypropylene indicator. The gauge has a maximum temperature rating of 250°F and a maximum pressure rating of 150 psi. It has a 4 inch dial with a dual scale calibrated in inches and centimeters. The gauge provides continuous monitoring of liquid level in tanks or vessels. It can be mounted either vertically or horizontally. This industrial liquid level gauge is made in the USA by Krueger Sentry Gauge.