Learn More
Grainger G3269 Level Gauge 304 Stainless Steel D 85in
Catalog No. 502767367
Encompass_Preferred
The Krueger 16N467 is a liquid level gauge designed for use with hydraulic reservoirs and gear boxes. It features a polycarbonate lens and aluminum housing for durability. The gauge has a dual scale that reads in both inches and millimeters with a range of 0-8 inches/0-200mm. It uses a bi-color indicator to clearly show the fluid level. This mechanical float gauge can withstand operating temperatures between -40 and 250°F. It has a 1/2" NPT connection and comes with a mounting gasket. The 16N467 provides at-a-glance monitoring of fluid levels.
Catalog No. 50-276-7367 Supplier Grainger Supplier No. 16N467
The Krueger 16N467 is a liquid level gauge designed for use with hydraulic reservoirs and gear boxes. It features a polycarbonate lens and aluminum housing for durability. The gauge has a dual scale that reads in both inches and millimeters with a range of 0-8 inches/0-200mm. It uses a bi-color indicator to clearly show the fluid level. This mechanical float gauge can withstand operating temperatures between -40 and 250°F. It has a 1/2" NPT connection and comes with a mounting gasket. The 16N467 provides at-a-glance monitoring of fluid levels.