Learn More
Grainger G3269 Level Gauge 304 Stainless Steel D 73in
Catalog No. 502765133
Encompass_Preferred
The Krueger 73-inch Level Gauge is a float-operated liquid level gauge designed for use with 73-inch containers. It features a hermetically sealed reed switch that is activated by a float to provide level indication. The gauge has an adjustable stem that allows the switch actuation point to be set at any liquid level. It can handle operating temperatures from -40 to 257°F and pressures up to 500 psi. The gauge has a stainless steel stem and float, with brass and stainless steel fittings. It comes with a 73-inch stem length and 3/4 inch NPT connections. This heavy duty industrial gauge is made in the USA by Krueger Sentry Gauge.
Catalog No. 50-276-5133 Supplier Grainger Supplier No. 16N456
The Krueger 73-inch Level Gauge is a float-operated liquid level gauge designed for use with 73-inch containers. It features a hermetically sealed reed switch that is activated by a float to provide level indication. The gauge has an adjustable stem that allows the switch actuation point to be set at any liquid level. It can handle operating temperatures from -40 to 257°F and pressures up to 500 psi. The gauge has a stainless steel stem and float, with brass and stainless steel fittings. It comes with a 73-inch stem length and 3/4 inch NPT connections. This heavy duty industrial gauge is made in the USA by Krueger Sentry Gauge.