Learn More
Grainger G3269 Level Gauge 304 Stainless Steel D 83in
Catalog No. 502765134
Encompass_Preferred
The Krueger Level Gauge For 83 in Container is a mechanical level gauge designed for use with 83 inch tall containers. It features a stainless steel body with a polypropylene float that moves up and down a center guide rod to indicate liquid level. The gauge has a 6 inch scale and is adjustable to fit containers from 80 to 86 inches tall. It can handle temperatures up to 180°F and pressures up to 100 PSI. This heavy duty mechanical gauge provides accurate level indication without any power required.
Catalog No. 50-276-5134 Supplier Grainger Supplier No. 16N466
The Krueger Level Gauge For 83 in Container is a mechanical level gauge designed for use with 83 inch tall containers. It features a stainless steel body with a polypropylene float that moves up and down a center guide rod to indicate liquid level. The gauge has a 6 inch scale and is adjustable to fit containers from 80 to 86 inches tall. It can handle temperatures up to 180°F and pressures up to 100 PSI. This heavy duty mechanical gauge provides accurate level indication without any power required.