Learn More
Grainger G3269 Level Gauge 304 Stainless Steel D 58in
Catalog No. 502765175
Encompass_Preferred
The Krueger 16N444 is a liquid level gauge designed for use with hydraulic reservoirs and gear boxes. It features a polycarbonate lens and aluminum housing for durability. The gauge has a dual scale that reads in US gallons and liters with a range of 0-15 gallons/0-60 liters. It uses a float and microswitch to detect fluid level and actuate warning lights. Key features include a green "normal" and red "low level" warning light, 1/2" NPT bottom connection, and it can withstand operating temperatures from -40°F to 250°F. This rugged and versatile level gauge is ideal for industrial machinery that requires fluid level monitoring.
Catalog No. 50-276-5175 Supplier Grainger Supplier No. 16N444
The Krueger 16N444 is a liquid level gauge designed for use with hydraulic reservoirs and gear boxes. It features a polycarbonate lens and aluminum housing for durability. The gauge has a dual scale that reads in US gallons and liters with a range of 0-15 gallons/0-60 liters. It uses a float and microswitch to detect fluid level and actuate warning lights. Key features include a green "normal" and red "low level" warning light, 1/2" NPT bottom connection, and it can withstand operating temperatures from -40°F to 250°F. This rugged and versatile level gauge is ideal for industrial machinery that requires fluid level monitoring.