Learn More
Grainger G3269 Level Gauge 304 Stainless Steel D 49in
Catalog No. 502765394
Encompass_Preferred
The Krueger Level Gauge For 49 in Container is a mechanical level gauge designed for 49 inch high containers. It features a stainless steel body and polypropylene float to provide chemical resistance. The gauge has a hermetically sealed design to prevent leakage and uses a magnetic coupling system to transmit the float position outside of the container. This allows visual monitoring of liquid level through the site glass without risk of leakage. The gauge mounts to the top of the container and is adjustable to fit different container diameters from 22 to 30 inches. It has a maximum temperature rating of 250°F.
Catalog No. 50-276-5394 Supplier Grainger Supplier No. 16N435
The Krueger Level Gauge For 49 in Container is a mechanical level gauge designed for 49 inch high containers. It features a stainless steel body and polypropylene float to provide chemical resistance. The gauge has a hermetically sealed design to prevent leakage and uses a magnetic coupling system to transmit the float position outside of the container. This allows visual monitoring of liquid level through the site glass without risk of leakage. The gauge mounts to the top of the container and is adjustable to fit different container diameters from 22 to 30 inches. It has a maximum temperature rating of 250°F.