Learn More
Grainger G3269 Level Gauge 304 Stainless Steel D 16in
Catalog No. 502764070
Encompass_Preferred
The Krueger 16N408 is a liquid level gauge designed for use with hydraulic reservoirs and gear boxes. It features a cast aluminum body with a clear acrylic lens for viewing fluid level. The gauge has a dual scale that reads in inches and centimeters with a max reading of 8 inches or 20 cm. It has a 1/2 inch NPT bottom mount thread and includes a locknut for easy installation. The gauge can withstand operating temperatures from -40 to 250°F. Overall it provides a simple visual method to monitor fluid levels in machinery.
Catalog No. 50-276-4070 Supplier Grainger Supplier No. 16N408
The Krueger 16N408 is a liquid level gauge designed for use with hydraulic reservoirs and gear boxes. It features a cast aluminum body with a clear acrylic lens for viewing fluid level. The gauge has a dual scale that reads in inches and centimeters with a max reading of 8 inches or 20 cm. It has a 1/2 inch NPT bottom mount thread and includes a locknut for easy installation. The gauge can withstand operating temperatures from -40 to 250°F. Overall it provides a simple visual method to monitor fluid levels in machinery.