Learn More
Grainger G3269 Level Gauge 304 Stainless Steel D 97in
Catalog No. 502765581
Encompass_Preferred
The Krueger 16N478 is a liquid level gauge designed for use with hydraulic reservoirs and gear boxes. It features a clear acrylic window with green and red zones for quick visual monitoring of oil level. The gauge has a brass body and is compatible with SAE 10, 20, 30, and 40 weight oils. It has a maximum pressure rating of 100 PSI and temperature range of -40°F to 250°F. The gauge has 1/2" NPT threads for easy installation and its compact 2-1/4" design allows it to fit well in tight spaces. Overall, this is a heavy-duty, industrial-grade level gauge for hydraulic fluid level indication.
Catalog No. 50-276-5581 Supplier Grainger Supplier No. 16N478
The Krueger 16N478 is a liquid level gauge designed for use with hydraulic reservoirs and gear boxes. It features a clear acrylic window with green and red zones for quick visual monitoring of oil level. The gauge has a brass body and is compatible with SAE 10, 20, 30, and 40 weight oils. It has a maximum pressure rating of 100 PSI and temperature range of -40°F to 250°F. The gauge has 1/2" NPT threads for easy installation and its compact 2-1/4" design allows it to fit well in tight spaces. Overall, this is a heavy-duty, industrial-grade level gauge for hydraulic fluid level indication.